Proteintech
Manufacturer Code:600061IG150UL
Catalog # 50-173-6188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C6G4 |
Immunogen | CD38 Fusion Protein Ag0487 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADP ribosyl cyclase 1 cADPr hydrolase 1 CD38 CD38 molecule Cyclic ADP ribose hydrolase 1 T10; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; ADPRC 1; cADPr hydrolase 1; CD38; CD38 antigen (p45); cluster of differentiation 38; Cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NIM-R5 antigen; T10
Gene Aliases: ADPRC 1; ADPRC1; CD38; Cd38-rs1
UniProt ID: (Human) P28907, (Mouse) P56528
Entrez Gene ID: (Human) 952, (Mouse) 12494
Molecular Function:
cyclase
glycosidase
hydrolase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.