R&D Systems
Manufacturer Code:FAB4625P
Catalog # FAB4625P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 424925 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CD37 Accession # NP_001765 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD37; CD37 CD37 antigentetraspanin-26 CD37 molecule cell differentiation antigen 37 GP52-40 leukocyte antigen CD37 MGC120234 Tetraspanin-26 Tspan-26 TSPAN26 TSPAN26leukocyte surface antigen CD37; cell differentiation antigen 37; Leukocyte antigen CD37; leukocyte surface antigen CD37; Tetraspanin-26; Tspan-26
Gene Aliases: CD37; GP52-40; TSPAN26
UniProt ID: (Human) P11049
Entrez Gene ID: (Human) 951
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.