Proteintech
Manufacturer Code:210441AP150UL
Catalog # 50-173-3225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD37 Fusion Protein Ag15384 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD37; CD37 antigen; CD37 CD37 molecule GP52 40 Leukocyte antigen CD37 Tetraspanin 26 Tspan 26 TSPAN26; cell differentiation antigen 37; Leukocyte antigen CD37; leukocyte surface antigen CD37; Tetraspanin-26; Tspan-26
Gene Aliases: CD37; GP52-40; TSPAN26
UniProt ID: (Human) P11049, (Mouse) Q61470, (Rat) P31053
Entrez Gene ID: (Human) 951, (Mouse) 12493, (Rat) 29185
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.