Invitrogen
Manufacturer Code:13121981
Catalog # 50-117-26
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBio4A9 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD360; IL-21 receptor; IL-21R; Interleukin-21 Receptor; LR-beta; Lymphocyte receptor beta; Novel cytokine receptor 8; Novel interleukin receptor; NR8
Gene Aliases: Il21r; NILR
UniProt ID: (Mouse) Q9JHX3
Entrez Gene ID: (Mouse) 60504
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.