Invitrogen
Manufacturer Code:17355941
Catalog # 17-355-941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Cr24.1 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD355; CD355 class I MHC restricted T cell associated molecule class-I MHC-restricted T cell associated molecule class-I MHC-restricted T-cell-associated molecule cytotoxic and regulatory T cell molecule; class I MHC restricted T cell associated molecule; class-I MHC-restricted T cell associated molecule; Class-I MHC-restricted T-cell-associated molecule; cytotoxic and regulatory T cell molecule; Cytotoxic and regulatory T-cell molecule
Gene Aliases: CD355; CRTAM
UniProt ID: (Human) O95727
Entrez Gene ID: (Human) 56253
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.