Novus Biologicals
Manufacturer Code:NBP2445670.2MG
Catalog # NBP24456702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | QBEnd/10 + HPCA1/763 |
Immunogen | Detergent solubilized vesicular suspension prepared from human term placenta (QBEnd/10) Recombinant full-length human HPCA1 protein (HPCA1/763) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD34; CD34 antigen; CD34 antigenhematopoietic progenitor cell antigen CD34 CD34 molecule; Hematopoietic progenitor cell antigen CD34
Gene Aliases: CD34
UniProt ID: (Human) P28906
Entrez Gene ID: (Human) 947
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.