Invitrogen
Manufacturer Code:PA578978
Catalog # PIPA578978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human CD34. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD34; CD34 antigen; cluster designation 34; Hematopoietic progenitor cell antigen CD34; Hematopoietic progenitor cell antigen CD34 CD34 CD34
Gene Aliases: AU040960; CD34
UniProt ID: (Human) P28906, (Mouse) Q64314
Entrez Gene ID: (Human) 947, (Rat) 305081, (Mouse) 12490
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.