Novus Biologicals
Manufacturer Code:NBP298385AF488
Catalog # NB082365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant CD320/TCblR/8D6A (NP_062294.3 Met1-Gly208). |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8D6 antigen; 8D6 antigen 8D6AFDC-SM-8D6 CD320 antigenFDC-signaling molecule 8D68D6TCBLR CD320 molecule TCblR Transcobalamin receptor; CD320; CD320 antigen; FDC-signaling molecule 8D6; FDC-SM-8D6; TCblR; Transcobalamin receptor
Gene Aliases: 8D6; 8D6A; CD320; TCBLR; UNQ198/PRO224
UniProt ID: (Human) Q9NPF0
Entrez Gene ID: (Human) 51293
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.