Invitrogen
Manufacturer Code:16587238
Catalog # 16-587-238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | A10 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: CD314; CD314 D12S2489E D6H12S2489E killer cell lectin like receptor K1 Killer cell lectin-like receptor subfamily K member 1 killer cell lectin-like receptor subfamily K member 1 KLR Klrk1 natural killer cell group 2D natural killer cell receptor NK cell receptor D NK lectin-like receptor Nkg2d NKG2-D NKG2-D type II integral membrane protein NKG2-D-activating NK receptor NKLLR Nkrp2 NKR-P2 NKR-P2 ortholog of human NKG2D; Killer cell lectin-like receptor subfamily K member 1; natural killer cell group 2D; NK cell receptor D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor
Gene Aliases: D6H12S2489E; Klrk1; NKG2-D; Nkg2d
UniProt ID: (Mouse) O54709
Entrez Gene ID: (Mouse) 27007
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.