Invitrogen
Manufacturer Code:17587842
Catalog # 50-112-3151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D11 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD314; CD314 NK cell receptor D NKG2-D type II integral membrane protein NKG2-D-activating NK receptor killer cell lectin-like receptor subfamily K member 1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; NK cell receptor D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor
Gene Aliases: CD314; D12S2489E; KLR; KLRK1; NKG2-D; NKG2D
UniProt ID: (Human) P26718
Entrez Gene ID: (Human) 22914
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.