Invitrogen
Manufacturer Code:14304280
Catalog # 14-304-280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | XML1Y1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: A5 protein; A5 protein BDCA4 C530029I03 CD304 Neuropilin neuropilin 1 Neuropilin1 neuropilin-1 Neuropilin-1 precursor (A5 protein) NP1 NP-1 Npn1 NPN-1 NRP NRP 1 Nrp1 sNRP 1 sNRP1 soluble NRP 1 soluble NRP1 transmembrane receptor vascular endothelial cell growth factor 165 receptor VEGF165R; CD304; Neuropilin-1; Neuropilin-1 precursor (A5 protein); transmembrane receptor; Vascular endothelial cell growth factor 165 receptor
Gene Aliases: BDCA4; C530029I03; CD304; NP-1; NP1; NPN-1; Npn1; NRP; NRP1; VEGF165R
UniProt ID: (Human) O14786, (Mouse) P97333
Entrez Gene ID: (Human) 8829, (Mouse) 18186
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.