R&D Systems
Manufacturer Code:MAB2640
Catalog # MAB2640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 232612 |
Immunogen | Y3 rat myeloid cell line transfected with human CD300a/LMIR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD300 antigen-like family member A; CD300a; CD300a antigen; CD300a CD300a antigenCMRF35H9 CD300a molecule CLM8 CLM-8 CMRF-35H CMRF35-H CMRF35H leukocyte immunoglobulin-like receptor CMRF35-H9 CMRF-35-H9CD300 antigen-like family member A CMRF35HIRp60 CMRF35-like molecule 8 IgSF12 IGSF12NK inhibitory receptor Immunoglobulin superfamily member 12 Inhibitory receptor protein 60 IRC1 IRC1/IRC2 IRC2 IRp60 leukocyte membrane antigen LMIR1 MAIR-I Mcipr1 NKRL Pigr4; CLM-8; CMRF-35-H9; CMRF35-H; CMRF35-H9; CMRF35-like molecule 8; CMRF35H leukocyte immunoglobulin-like receptor; IgSF12; Immunoglobulin superfamily member 12; Inhibitory receptor protein 60; IRC1/IRC2; IRp60; leukocyte membrane antigen; NK inhibitory receptor
Gene Aliases: CD300A; CLM-8; CMRF-35-H9; CMRF-35H; CMRF35-H; CMRF35-H9; CMRF35H; CMRF35H9; HSPC083; IGSF12; IRC1; IRC1/IRC2; IRC2; IRp60
UniProt ID: (Human) Q9UGN4
Entrez Gene ID: (Human) 11314
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.