Invitrogen
Manufacturer Code:MA523927
Catalog # PIMA523927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 172224 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse CD300a/LMIR1 Leu28-Arg183 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD300 antigen-like family member A; CD300a; CD300a CLM8 CMRF-35H IGSF12 IRC1 IRC2 IRp60 LMIR1 MAIR-I Mcipr1 NKRL Pigr4 CD300a antigenCMRF35H9 CD300a molecule CLM-8 CMRF-35H CMRF35-H CMRF35H leukocyte immunoglobulin-like receptor CMRF35-H9 CMRF-35-H9CD300 antigen-like family member A CMRF35HIRp60 CMRF35-like molecule 8 IgSF12 IGSF12NK inhibitory receptor Immunoglobulin superfamily member 12 Inhibitory receptor protein 60 IRC1 IRC1/IRC2 IRC2 Irp60 leukocyte membrane antigen; CLM-8; CMRF35-like molecule 8; Leukocyte mono-Ig-like receptor 1; MAIR-1; Mast cell-derived paired immunoglobulin-like receptor 1; Myeloid-associated immunoglobulin-like receptor 1; polymeric immunoglobulin receptor 4
Gene Aliases: B230315M08Rik; Cd300a; Clm8; LMIR1; MAIR-I; MAIR-Ia; Mcpir1; MMAC8; Pigr4
UniProt ID: (Mouse) Q6SJQ0
Entrez Gene ID: (Mouse) 217303
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.