Invitrogen
Manufacturer Code:PA5113473
Catalog # PIPA5113473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human CMRF35-like molecule 8 protein (18-180AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B230315M08Rik CD300 antigen-like family member A CD300A CD300a antigen CD300a molecule Clm8 CLM-8 CMRF35H CMRF-35H CMRF35-H CMRF35H leukocyte immunoglobulin-like receptor CMRF35H9 CMRF-35-H9 CMRF35-H9 CMRF35HIRp60 CMRF35-like molecule 8 HSPC083 IGSF12 Immunoglobulin superfamily member 12 Inhibitory receptor protein 60 IRC1 IRC1/IRC2 IRC2 IRp60 leukocyte membrane antigen Leukocyte mono-Ig-like receptor 1 Lmir1 MAIR-1 MAIR-I MAIR-Ia Mast cell-derived paired immunoglobulin-like receptor 1 Mcipr1 Mcpir1 MMAC8 Myeloid-associated immunoglobulin-like receptor 1 NK inhibitory receptor NKRL Pigr4 polymeric immunoglobulin receptor 4; CD300 antigen-like family member A; CD300a; CD300a antigen; CLM-8; CMRF-35-H9; CMRF35-H; CMRF35-H9; CMRF35-like molecule 8; CMRF35H leukocyte immunoglobulin-like receptor; IgSF12; Immunoglobulin superfamily member 12; Inhibitory receptor protein 60; IRC1/IRC2; IRp60; leukocyte membrane antigen; Leukocyte mono-Ig-like receptor 1; MAIR-1; Mast cell-derived paired immunoglobulin-like receptor 1; Myeloid-associated immunoglobulin-like receptor 1; NK inhibitory receptor; polymeric immunoglobulin receptor 4
Gene Aliases: B230315M08Rik; CD300A; CLM-8; Clm8; CMRF-35-H9; CMRF-35H; CMRF35-H; CMRF35-H9; CMRF35H; CMRF35H9; HSPC083; IGSF12; IRC1; IRC1/IRC2; IRC2; IRp60; LMIR1; MAIR-I; MAIR-Ia; Mcpir1; MMAC8; Pigr4
UniProt ID: (Human) Q9UGN4, (Mouse) Q6SJQ0
Entrez Gene ID: (Human) 11314, (Mouse) 217303
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.