Abnova
Manufacturer Code:PAB29578
Catalog # 89-933-778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 98-149 of human CD300A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant human CD300A.
For Research Use Only
Protein Aliases: CD300 antigen-like family member A; CD300a; CD300a antigen; CLM-8; CMRF-35-H9; CMRF-35H; CMRF35-H; CMRF35-H9; CMRF35-like molecule 8; CMRF35H; CMRF35H leukocyte immunoglobulin-like receptor; CMRF35H9; IGSF12; Immunoglobulin superfamily member 12; Inhibitory receptor protein 60; IRC1; IRC1/IRC2; IRC2; IRp60; leukocyte membrane antigen; NK inhibitory receptor
Gene Aliases: CD300A; CLM-8; CMRF-35-H9; CMRF-35H; CMRF35-H; CMRF35-H9; CMRF35H; CMRF35H9; HSPC083; IGSF12; IRC1; IRC1/IRC2; IRC2; IRp60
UniProt ID: (Human) Q9UGN4
Entrez Gene ID: (Human) 11314
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.