Novus Biologicals
Manufacturer Code:NBP232636
Catalog # NBP232636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: GQDGVRQSRASDKQTLLPNDQLYQPLKDREDDQYSHLQGNQLRRN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD3g; CD3g antigen CD3g antigen gamma polypeptide (TiT3 complex) CD3g molecule epsilon (CD3-TCR complex) CD3g molecule gamma (CD3-TCR complex) CD3-GAMMA FLJ17620 FLJ17664 FLJ79544 FLJ94613 MGC138597 T3G T-cell antigen receptor complex gamma subunit of T3 T-cell receptor T3 gamma chain T-cell surface glycoprotein CD3 gamma chain; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain
Gene Aliases: CD3-GAMMA; CD3G; IMD17; T3G
UniProt ID: (Human) P09693
Entrez Gene ID: (Human) 917
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.