Novus Biologicals
Manufacturer Code:NBP224867
Catalog # NBP224867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunoblot (IB) | Assay dependent |
In vitro Assay (IV) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OKT3 |
Immunogen | Human PBMCs were used as immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: anti-CD3 okt3 CD3 antigen delta subunit CD3d antigen CD3d antigen delta polypeptide (TiT3 complex) CD3d molecule delta (CD3-TCR complex) CD3-DELTA CD3e antigen CD3e antigen epsilon polypeptide (TiT3 complex) CD3e molecule epsilon (CD3-TCR complex) CD3-epsilon CD3G CD3g antigen CD3g antigen gamma polypeptide (TiT3 complex) CD3g molecule epsilon (CD3-TCR complex) CD3g molecule gamma (CD3-TCR complex) CD3-GAMMA FLJ17620 FLJ17664 FLJ18683 FLJ79544 FLJ94613 MGC138597 okt3 okt3 ab okt3 CD3 okt3 hybridoma T3DOKT3 delta chain T3E T-cell antigen receptor complex epsilon subunit of T3 T-cell receptor T3 delta chain T-cell surface antigen T3/Leu-4 epsilon chain T-cell surface glycoprotein CD3 delta chain T-cell surface glycoprotein CD3 epsilon chain TCRE; CD3-epsilon; CD3e; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule, epsilon (CD3-TCR complex); T-cell antigen receptor complex, epsilon subunit of T3; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 epsilon chain
Gene Aliases: CD3E; IMD18; T3E; TCRE
UniProt ID: (Human) P07766
Entrez Gene ID: (Human) 916
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.