Invitrogen
Manufacturer Code:PA559603
Catalog # PIPA559603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CD2BP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD2 (cytoplasmic tail) binding protein 2; CD2 antigen (cytoplasmic tail) binding protein 2; CD2 antigen cytoplasmic tail-binding protein 2; CD2 cytoplasmic domain-binding protein 2; CD2BP2 FWP010 LIN1 Snu40 CD2 antigen cytoplasmic tail binding protein 2 CD2 cytoplasmic tail binding protein 2 CD2 antigen cytoplasmic tail-binding protein 2 U5-52K CD2 binding protein 2 CD2 cytoplasmic domain binding protein 2 U5 snRNP 52K protein KIAA1178; protein phosphatase 1, regulatory subunit 59; U5 snRNP 52K protein; U5-52K
Gene Aliases: CD2BP2; FWP010; KIAA1178; LIN1; PPP1R59; Snu40; U5-52K
UniProt ID: (Human) O95400
Entrez Gene ID: (Human) 10421
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.