Invitrogen
Manufacturer Code:MA533009
Catalog # PIMA533009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F8 |
Immunogen | E.coli-derived human CD2AP recombit protein (Position: K253-K337). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adapter protein CMS; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2-associated protein; CD2-associated protein Adapter protein CMS Cas ligand with multiple SH3 domains CD2AP; mesenchyme to epithelium transition protein, SH3 domains; Mesenchyme-to-epithelium transition protein with SH3 domains 1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1
Gene Aliases: AL024079; C78928; CD2AP; CMS; METS-1; Mets1
UniProt ID: (Human) Q9Y5K6, (Mouse) Q9JLQ0
Entrez Gene ID: (Human) 23607, (Mouse) 12488, (Rat) 316258
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.