Invitrogen
Manufacturer Code:11902282
Catalog # 50-100-60
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | mT2.7 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD282; TIL4 Toll-like Receptor 2 toll/interleukin 1 receptor-like 4 toll/interleukin-1 receptor-like protein 4; Toll-like receptor 2
Gene Aliases: Ly105; Tlr2
UniProt ID: (Mouse) Q9QUN7
Entrez Gene ID: (Mouse) 24088
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.