Invitrogen
Manufacturer Code:16992282
Catalog # 50-112-3015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TL2.1 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD282; TIL4 Toll-like Receptor 2 toll/interleukin 1 receptor-like 4 toll/interleukin-1 receptor-like protein 4; Toll-like receptor 2; Toll/interleukin-1 receptor-like protein 4
Gene Aliases: CD282; TIL4; TLR2
UniProt ID: (Human) O60603
Entrez Gene ID: (Human) 7097
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.