Invitrogen
Manufacturer Code:14901195
Catalog # 14-901-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBioTR23 (TR23) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD281; CD281 KIAA0012 rsc786 TIL TIL. LPRS5 Tlr1 toll like receptor 1 Toll/interleukin-1 receptor-like protein toll-like receptor 1; TIL; Toll-like receptor 1; Toll/interleukin-1 receptor-like protein
Gene Aliases: Tlr1
UniProt ID: (Mouse) Q9EPQ1
Entrez Gene ID: (Mouse) 21897
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.