Novus Biologicals
Manufacturer Code:NBP127932
Catalog # NBP127932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | PV-1 |
Immunogen | C57BL/6N mouse T cell lymphoma EL4 cell line |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: CD28; CD28 antigen; CD28 antigen CD28 antigen (Tp44) CD28 molecule MGC138290 T-cell-specific surface glycoprotein CD28 Tp44; T-cell-specific surface glycoprotein CD28; TP44
Gene Aliases: CD28; Tp44
UniProt ID: (Human) P10747
Entrez Gene ID: (Human) 940
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.