Invitrogen
Manufacturer Code:160280050
Catalog # 16-028-0050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | JJ319 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen CD28 Cd28 CD28 antigen CD28 antigen (Tp44) CD28 isoform CD28 isoform 2 CD28 molecule CD28 precursor protein CD28 protein CD28 protein precursor CD28RNA cell surface protein CHT28 contains partial extracellular domain costimulatory molecule B7 receptor CD28 EGK_04709 MGC138290 sCD28 soluble CD28 T44 T-cell costimulatory molecule CD28 T-cell costimulatory molecule Tp44 T-cell-specific surface glycoprotein CD28 T-cell-specific surface glycoprotein CD28 homolog TP44; CD28; CD28 antigen; T-cell-specific surface glycoprotein CD28
Gene Aliases: Cd28; CD28RNA
UniProt ID: (Rat) P31042
Entrez Gene ID: (Rat) 25660
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.