R&D Systems
Manufacturer Code:MAB342500
Catalog # MAB342500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 37407 |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line <em>Sf</em> 21-derived recombinant human CD28 Asn19-Pro152 Accession # P10747 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD28; CD28 antigen; CD28 CD28 antigen CD28 antigen (Tp44) CD28 molecule MGC138290 T-cell-specific surface glycoprotein CD28 Tp44; T-cell-specific surface glycoprotein CD28; TP44
Gene Aliases: CD28; Tp44
UniProt ID: (Human) P10747
Entrez Gene ID: (Human) 940
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.