Invitrogen
Manufacturer Code:45028180
Catalog # 50-157-86
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Syrian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 37.51 |
Conjugate | PerCP-Cyanine5.5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Syrian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: CD28; MGC138290 Tp44 CD28 antigen T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28
Gene Aliases: Cd28
UniProt ID: (Mouse) P31041
Entrez Gene ID: (Mouse) 12487
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.