Invitrogen
Manufacturer Code:14661995
Catalog # 14-661-995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MIH24 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD254; CD254 H-RANK-L hRANKL2 Ly109l M-Rank-L ODF OPG OPG ligand OPGL OPTB2 Osteoclast differentiation factor osteoprotegerin ligand Rankl rank-l receptor activator of NF-kappaB ligand receptor activator of nuclear factor kappa B ligand receptor activator of nuclear factor kappa-B ligand RP11-86N24.2 sOdf srankl TNF-related activation-induced cytokine Tnfsf11 TNLG6B Trance tumor necrosis factor (ligand) superfamily member 11 tumor necrosis factor (ligand) superfamily member 11 tumor necrosis factor ligand 6B tumor necrosis factor ligand superfamily member 11 Tumor necrosis factor ligand superfamily member 11 membrane form Tumor necrosis factor ligand superfamily member 11 soluble form tumor necrosis factor superfamily member 11 tumor necrosis factor-related activation-induced cytokine; ODF; OPGL; Osteoclast differentiation factor; Osteoprotegerin ligand; RANKL; receptor activator of nuclear factor kappa B ligand; Receptor activator of nuclear factor kappa-B ligand; TNF-related activation-induced cytokine; TRANCE; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; Tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form
Gene Aliases: CD254; hRANKL2; ODF; OPGL; OPTB2; RANKL; sOdf; TNFSF11; TNLG6B; TRANCE
UniProt ID: (Human) O14788
Entrez Gene ID: (Human) 8600
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.