Invitrogen
Manufacturer Code:12595281
Catalog # 50-110-13
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | IK22/5 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD254; ODF; OPG ligand; OPGL; Osteoclast differentiation factor; Osteoprotegerin ligand; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; Receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2 CD254 ODF OPGL OPTB2 RANKL TRANCE hRANKL2 sOdf TNF-related activation-induced cytokine TNFSF11 TRANCE; TNF-related activation-induced cytokine; TRANCE; Tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor-related activation-induced cytokine
Gene Aliases: Ly109l; ODF; OPG; OPGL; RANKL; Tnfsf11; Trance
UniProt ID: (Mouse) O35235
Entrez Gene ID: (Mouse) 21943
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.