Invitrogen
Manufacturer Code:PA520291
Catalog # PIPA520291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide from near the center of human sRANK-L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD254; ODF; OPG ligand; OPGL; Osteoclast differentiation factor; Osteoprotegerin ligand; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; Receptor activator of nuclear factor kappa-B ligand; RP11-86N24.2 CD254 ODF OPGL OPTB2 RANKL TRANCE hRANKL2 sOdf TNF-related activation-induced cytokine TNFSF11 TRANCE; TNF-related activation-induced cytokine; TRANCE; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; Tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor-related activation-induced cytokine
Gene Aliases: CD254; hRANKL2; Ly109l; ODF; OPG; OPGL; OPTB2; RANKL; sOdf; TNFSF11; TNLG6B; TRANCE
UniProt ID: (Human) O14788, (Rat) Q9ESE2, (Mouse) O35235
Entrez Gene ID: (Human) 8600, (Rat) 117516, (Mouse) 21943
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.