Invitrogen
Manufacturer Code:53025280
Catalog # 50-166-94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBio7D4 (7D4) |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD25; IL-2 receptor subunit alpha; IL-2-RA; IL-2R alpha chain; IL-2R subunit alpha; IL2 RA antibody Il2ra antibody Interleukin 2 receptor alpha chain antibody TAC antigen antibody IL-2R alpha chain; IL2-RA; Interleukin-2 receptor subunit alpha; p55; p55 chain
Gene Aliases: CD25; Il2r; Il2ra; Ly-43
UniProt ID: (Mouse) P01590
Entrez Gene ID: (Mouse) 16184
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.