Invitrogen
Manufacturer Code:MA180632
Catalog # PIMA180632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NDS 61 |
Immunogen | Rat interleukin-2 receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD25; IL-2 receptor alpha subunit; IL-2 receptor subunit alpha; IL-2-RA; IL-2R subunit alpha; IL2 RA antibody Il2ra antibody Interleukin 2 receptor alpha chain antibody TAC antigen antibody IL-2R alpha chain; IL2-RA; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha chain; Interleukin-2 receptor subunit alpha
Gene Aliases: Il2ra; IL2RAC
UniProt ID: (Rat) P26897
Entrez Gene ID: (Rat) 25704
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.