Invitrogen
Manufacturer Code:35583841
Catalog # 35-583-841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | eBioC1.7 (C1.7) |
Conjugate | PE-Cyanine5.5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2B4 NAIL NKR2B4 Nmrk SLAMF4 NK cell activation inducing ligand NAIL NK cell activation-inducing ligand NK cell type I receptor protein 2B4 h2B4 natural killer cell receptor 2B4; CD244; CD244 molecule, natural killer cell receptor 2B4; h2B4; NAIL; Natural killer cell receptor 2B4; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell type I receptor protein 2B4; NKR2B4; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4
Gene Aliases: 2B4; CD244; NAIL; NKR2B4; Nmrk; SLAMF4
UniProt ID: (Human) Q9BZW8
Entrez Gene ID: (Human) 51744
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily kinase protein kinase receptor signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.