Invitrogen
Manufacturer Code:16583838
Catalog # 16-583-838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | eBioC1.7 (C1.7) |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2B4 C9.1 Cd244 CD244 long isoform CD244 molecule Cd244 molecule natural killer cell receptor 2B4 CD244 natural killer cell receptor 2B4 CD244 short isoform F730046O15Rik h2B4 Ly90 NAIL Natural killer cell receptor 2B4 NK cell activation inducing ligand NAIL NK cell activation-inducing ligand NK cell receptor 2B4 NK cell type I receptor protein 2B4 NKR Nmrk NKR2B4 Nmrk non MHC restricted killing associated Non-MHC restricted killing associated signaling lymphocytic activation molecule 4 SLAM family member 4 SLAMF4 transmembrane NK cell receptor 2B4; CD244; CD244 molecule, natural killer cell receptor 2B4; h2B4; NAIL; Natural killer cell receptor 2B4; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell type I receptor protein 2B4; NKR2B4; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4
Gene Aliases: 2B4; CD244; NAIL; NKR2B4; Nmrk; SLAMF4
UniProt ID: (Human) Q9BZW8
Entrez Gene ID: (Human) 51744
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.