Abnova
Manufacturer Code:PAB17803
Catalog # 89-119-468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to internal region of residues surrounding Y266 of human CD244. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic phosphopeptide of CD244.
For Research Use Only
Protein Aliases: 2B4; CD244; CD244 molecule, natural killer cell receptor 2B4; h2B4; NAIL; Natural killer cell receptor 2B4; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell type I receptor protein 2B4; NKR2B4; Nmrk; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4
Gene Aliases: 2B4; CD244; NAIL; NKR2B4; Nmrk; SLAMF4
UniProt ID: (Human) Q9BZW8
Entrez Gene ID: (Human) 51744
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily kinase protein kinase receptor signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.