Invitrogen
Manufacturer Code:61024280
Catalog # 61-024-280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | M1/69 |
Conjugate | PE-eFluor 610 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD24; cluster of differentiation 24; heat stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; Nectadrin; R13-Ag; Signal transducer CD24; X62 heat stable antigen
Gene Aliases: Cd24; Cd24a; HSA; Ly-52; nectadrin
UniProt ID: (Mouse) P24807
Entrez Gene ID: (Mouse) 12484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.