R&D Systems
Manufacturer Code:FAB6900T025
Catalog # FAB6900T025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 691632 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CD23/Fc epsilon RII Glu50-Pro331 Accession # P20693 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BLAST-2; BLAST-2 CD23 CD23A CD23CD23 antigen CLEC4J CLEC4JC-type lectin domain family 4 member J C-type lectin domain family 4 member J Fc epsilon RII Fc fragment of IgE low affinity II receptor for (CD23) FCE2Fc fragment of IgE low affinity II receptor for (CD23A) fc-epsilon-RII FCER2 Fcer2a FceRII IGEBF Immunoglobulin E-binding factor low affinity immunoglobulin epsilon Fc receptor Ly-42 Lymphocyte IgE receptor; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; CD23; CD23 antigen; Fc epsilon receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc-epsilon-RII; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; Low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; Lymphocyte IgE receptor
Gene Aliases: BLAST-2; CD23; CD23A; CLEC4J; FCE2; FCER2; IGEBF
UniProt ID: (Human) P06734
Entrez Gene ID: (Human) 2208
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.