Invitrogen
Manufacturer Code:MA514572
Catalog # PIMA514572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP23 |
Immunogen | Recombit protein encoding human CD23 48-248aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BLAST-2; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; CD23; CD23 antigen; CD23 CD23A CLEC4J FCE2 IGEBF BLAST-2 C-type lectin domain family 4 member J CD23 antigen fc-epsilon-RII immunoglobulin E-binding factor low affinity immunoglobulin epsilon Fc receptor lymphocyte IgE receptor; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc-epsilon-RII; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; Low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Lymphocyte IgE receptor
Gene Aliases: BLAST-2; CD23; CD23A; CLEC4J; FCE2; FCER2; Fcer2a; IGEBF; Ly-42
UniProt ID: (Human) P06734, (Mouse) P20693
Entrez Gene ID: (Human) 2208, (Mouse) 14128
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.