Invitrogen
Manufacturer Code:12023282
Catalog # 50-112-2218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | B3B4 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD23; CD23 CD23A CLEC4J FCE2 IGEBF BLAST-2 C-type lectin domain family 4 member J CD23 antigen fc-epsilon-RII immunoglobulin E-binding factor low affinity immunoglobulin epsilon Fc receptor lymphocyte IgE receptor; FC epsilon RII; Fc-epsilon-RII; Low affinity immunoglobulin epsilon Fc receptor; low-affinity IgE receptor; Lymphocyte IgE receptor
Gene Aliases: CD23; Fce2; Fcer2; Fcer2a; Ly-42
UniProt ID: (Mouse) P20693
Entrez Gene ID: (Mouse) 14128
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.