Invitrogen
Manufacturer Code:MA517990
Catalog # PIMA517990
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 15F.10E5 |
Immunogen | AE7 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD226; CD226 antigen; DNAM-1 DNAM1 PTA1 TLiSA1 CD226 antigen DNAX accessory molecule 1 DNAX accessory molecule-1 T lineage-specific activation antigen 1 antigen adhesion glycoprotein platelet and T cell activation antigen 1; platelet and T cell activation antigen 1; Platelet and T-cell activation antigen 1
Gene Aliases: BC051526; Cd226; DNAM-1; DNAM1; Pta1; TLiSA1
UniProt ID: (Mouse) Q8K4F0
Entrez Gene ID: (Mouse) 225825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.