Invitrogen
Manufacturer Code:PA536393
Catalog # PIPA536393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 293-342 of human DNAM-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adhesion glycoprotein; adhesion glycoprotein CD226 antigen CD226 molecule DNAM 1 DNAM1 DNAX accessory molecule 1 platelet and T cell activation antigen 1 PTA1 T lineage specific activation antigen 1 antigen TLiSA1; CD226; CD226 antigen; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; Platelet and T-cell activation antigen 1; T lineage-specific activation antigen 1 antigen
Gene Aliases: BC051526; CD226; DNAM-1; DNAM1; PTA1; TLiSA1
UniProt ID: (Human) Q15762, (Mouse) Q8K4F0
Entrez Gene ID: (Human) 10666, (Rat) 307199, (Mouse) 225825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.