Abnova
Manufacturer Code:PAB26976
Catalog # 89-128-622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human CD226. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CD226.
For Research Use Only
Protein Aliases: adhesion glycoprotein; CD226; CD226 antigen; DNAM-1; DNAM1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; PTA1; T lineage-specific activation antigen 1 antigen; TLiSA1
Gene Aliases: CD226; DNAM-1; DNAM1; PTA1; TLiSA1
UniProt ID: (Human) Q15762
Entrez Gene ID: (Human) 10666
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.