Invitrogen
Manufacturer Code:14021195
Catalog # 14-021-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBio8D9 (8D9) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C3b/C4b receptor C3-binding protein C3DR CD21 CD35 CD35 antigen complement C3d receptor complement receptor 2 complement receptor type 1 complement receptor type 2 Cr1 Cr-1 Cr2 Cr-2 EBV receptor expressed in B lymphocytes; complement C3d receptor; complement receptor type 1; complement receptor type 2; expressed in B lymphocytes
Gene Aliases: C3DR; CD21; CD35; Cr-1; Cr-2; Cr1
Entrez Gene ID: (Mouse) 12902
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.