Invitrogen
Manufacturer Code:14209295
Catalog # 14-209-295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | LWC06 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD209; CD209 antigen-like protein A; CD209 CD209 antigen CD209 antigen-like protein A CD209 molecule Cd209a CD209a antigen CD209a molecule Cd209d CD209d antigen CDSIGN CIRE CLEC4L Clec4m C-type lectin domain family 4 member L C-type lectin domain family 4 member L C-type lectin domain family 4 member M Dcsign DC-SIGN DC-SIGN1 Dc-signr DC-SIGN-related protein Dendritic cell-specific ICAM-3-grabbing non-integrin dendritic cell-specific ICAM-3-grabbing non-integrin 1 dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin HIV gpl20-binding protein MGC129965 MGC130443 RGD1561104 SIGN-R1 SIGNR5; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin
Gene Aliases: CD209; Cd209a; CDSIGN; CIRE; DC-SIGN; DC-SIGN1; Dcsign; SIGN-R1; SIGNR5
UniProt ID: (Mouse) Q91ZX1
Entrez Gene ID: (Mouse) 170786
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.