Invitrogen
Manufacturer Code:PA532749
Catalog # PIPA532749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from internal region of human ENPP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alkaline phosphodiesterase I; B10 CD203c NPP3 PD-IBETA PDNP3 E-NPP 3; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); E-NPP 3; Ectonucleotide pyrophosphatase/phosphodiesterase family member 3; gp130RB13-6; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Phosphodiesterase I beta; Phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta
Gene Aliases: B10; CD203c; ENPP3; NPP3; PD-IBETA; PDNP3
UniProt ID: (Human) O14638
Entrez Gene ID: (Human) 5169
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.