Invitrogen
Manufacturer Code:MA517566
Catalog # PIMA517566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OX-2 |
Immunogen | Rat thymocyte membrane glycoproteins were fused with mouse NS-1 myeloma cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cd200 antigen; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); MRC OX-2 antigen; My033 MOX1 MOX2 MRC OX-2 CD200 antigen MRC OX-2 antigen OX-2 membrane glycoprotein antigen identified by monoclonal antibody MRC OX-2; OX-2 membrane glycoprotein
Gene Aliases: Cspmo2; Mox2; MRCOX2
Entrez Gene ID: (Rat) 24560
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.