Invitrogen
Manufacturer Code:MA523575
Catalog # PIMA523575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 325516 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human CD200 Gln31-Gly232 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen identified by monoclonal antibody MRC OX-2; CD200; CD200 antigen; My033 MOX1 MOX2 MRC OX-2 CD200 antigen MRC OX-2 antigen OX-2 membrane glycoprotein antigen identified by monoclonal antibody MRC OX-2; OX-2 membrane glycoprotein
Gene Aliases: CD200; MOX1; MOX2; MRC; My033; OX-2
UniProt ID: (Human) P41217
Entrez Gene ID: (Human) 4345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.