Proteintech
Manufacturer Code:140571AP150UL
Catalog # 50-172-9179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD200 Fusion Protein Ag5045 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen identified by monoclonal antibody MRC OX-2; CD200; CD200 antigen; CD200 CD200 molecule MOX1 MOX2 MRC OX 2 OX 2 membrane glycoprotein; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); MRC OX-2 antigen; OX-2 membrane glycoprotein
Gene Aliases: CD200; Cspmo2; MOX1; MOX2; MRC; MRCOX2; My033; OX-2; OX2
UniProt ID: (Human) P41217
Entrez Gene ID: (Human) 4345, (Mouse) 17470, (Rat) 24560
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.