R&D Systems
Manufacturer Code:MAB6979
Catalog # MAB6979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 703335 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CD1d Glu20-Ser301 (predicted) Accession # P15813 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antigen-presenting glycoprotein CD1d; antigen-presenting glycoprotein CD1d CD1A CD1d CD1d antigen CD1D antigen d polypeptide CD1d molecule differentiation antigen CD1-alpha-3 HMC class I antigen-like glycoprotein CD1D MGC34622 R3 R3G1 T-cell surface glycoprotein CD1d thymocyte antigen CD1D; CD1d; CD1D antigen, d polypeptide; differentiation antigen CD1-alpha-3; HMC class I antigen-like glycoprotein CD1D; R3G1; T-cell surface glycoprotein CD1d; thymocyte antigen CD1D
Gene Aliases: CD1A; CD1D; R3; R3G1
UniProt ID: (Human) P15813
Entrez Gene ID: (Human) 912
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.