Invitrogen
Manufacturer Code:13001185
Catalog # 50-113-61
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 1B1 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AI747460 CD1.1 Cd1a Cd1d Ly-38 T-cell surface glycoprotein CD1d1 antigen-presenting glycoprotein CD1d1; Antigen-presenting glycoprotein CD1d1; CD1d.1; T-cell surface glycoprotein CD1d1
Gene Aliases: AI747460; CD1.1; Cd1a; Cd1d; Cd1d1; Ly-38
UniProt ID: (Mouse) P11609
Entrez Gene ID: (Mouse) 12479
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.