Proteintech
Manufacturer Code:173361AP150UL
Catalog # 50-173-1516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD1d Fusion Protein Ag11371 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen-presenting glycoprotein CD1d; Antigen-presenting glycoprotein CD1d1; CD1A CD1D CD1d molecule R3 R3G1; CD1d; CD1D antigen, d polypeptide; CD1d.1; differentiation antigen CD1-alpha-3; HMC class I antigen-like glycoprotein CD1D; R3G1; T-cell surface glycoprotein CD1d; T-cell surface glycoprotein CD1d1; thymocyte antigen CD1D
Gene Aliases: AI747460; CD1.1; CD1A; CD1D; Cd1d1; Ly-38; R3; R3G1
UniProt ID: (Human) P15813, (Mouse) P11609
Entrez Gene ID: (Human) 912, (Mouse) 12479
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.